Skip to Content
Merck
All Photos(1)

Key Documents

52487

Supelco

Denatonium benzoate

PESTANAL®, analytical standard

Synonym(s):

N,N-Diethyl-N-[(2,6-dimethylphenyl­carbamoyl)­methyl]­benzyl­ammonium benzoate, Benzyldiethyl(2,6-xylyl­carbamoyl­methyl)­ammonium benzoate, THS 839

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H29N2O · C7H5O2
CAS Number:
Molecular Weight:
446.58
Beilstein:
8179408
EC Number:
MDL number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥97.0% (HPLC)
97.0-103.0% (NT)

shelf life

limited shelf life, expiry date on the label

mp

164-168 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[O-]C(=O)c1ccccc1.CC[N+](CC)(CC(=O)Nc2c(C)cccc2C)Cc3ccccc3

InChI

1S/C21H28N2O.C7H6O2/c1-5-23(6-2,15-19-13-8-7-9-14-19)16-20(24)22-21-17(3)11-10-12-18(21)4;8-7(9)6-4-2-1-3-5-6/h7-14H,5-6,15-16H2,1-4H3;1-5H,(H,8,9)

InChI key

VWTINHYPRWEBQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service