Skip to Content
Merck
All Photos(1)

Key Documents

45528

Supelco

4-Hydroxybenzonitrile

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCC6H4OH
CAS Number:
Molecular Weight:
119.12
Beilstein:
386130
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

110-113 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Oc1ccc(cc1)C#N

InChI

1S/C7H5NO/c8-5-6-1-3-7(9)4-2-6/h1-4,9H

InChI key

CVNOWLNNPYYEOH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Pirot et al.
Proceedings of the National Academy of Sciences of the United States of America, 94(4), 1562-1567 (1997-02-18)
Attenuated-total-reflectance Fourier-transform-infrared spectroscopy has been used to rapidly and noninvasively quantify in vivo the uptake of a chemical into the outermost, and least permeable, layer of human skin (the stratum corneum). The objective of the experiment was to develop a
N Higo et al.
Pharmaceutical research, 10(10), 1500-1506 (1993-10-01)
Attenuated total-reflectance infrared (ATR-IR) spectroscopy has been used to follow the penetration of a model compound (4-cyanophenol; CP) across human stratum corneum (SC) in vivo, in man. CP was administered for periods of 1, 2, or 3 hr, either (a)
V H Mak et al.
Pharmaceutical research, 7(8), 835-841 (1990-08-01)
A novel application of attenuated total reflectance IR spectroscopy (ATR-IR) was used to monitor the outer several microns of the stratum corneum (SC) and, thereby, demonstrate enhanced percutaneous absorption in vivo in man. 4-Cyanophenol (CP) as a model permeant yielded
A L Stinchcomb et al.
Pharmaceutical research, 16(8), 1288-1293 (1999-09-01)
Simple, safe and quick in vivo methods for estimating chemical uptake into the stratum corneum (SC) from volatile and non-volatile solvents are invaluable to health risk assessors. This study compares the human in vivo SC uptake of a model compound
J E Harrison et al.
Pharmaceutical research, 13(4), 542-546 (1996-04-01)
The purpose of this work was to analyse the mechanism of the enhancement of percutaneous penetration demonstrated by the known enhancers Azone and Transcutol. Enhancer induced changes in the diffusivity and solubility of a model permeant (4-cyanophenol) in human stratum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service