Skip to Content
Merck
All Photos(1)

Key Documents

41510

Sigma-Aldrich

2,2-Dimethyl-1,3-propanediol

purum, ≥98.0% (GC)

Synonym(s):

Neopentylglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(CH2OH)2
CAS Number:
Molecular Weight:
104.15
Beilstein:
605291
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3.6 (vs air)

vapor pressure

<0.8 mmHg ( 20 °C)

grade

purum

Assay

≥98.0% (GC)

autoignition temp.

750 °F

expl. lim.

1.34 %, 149 °F
18.8 %, 177 °F

mp

126-128 °C

SMILES string

CC(C)(CO)CO

InChI

1S/C5H12O2/c1-5(2,3-6)4-7/h6-7H,3-4H2,1-2H3

InChI key

SLCVBVWXLSEKPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Review

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B. Cornils et al.
Chem. Zeit., 100, 504-504 (1976)
Irimpan Mathews et al.
Biochemistry, 46(31), 8969-8979 (2007-07-20)
The unusual architecture of the enzyme (MsAcT) isolated from Mycobacterium smegmatis forms the mechanistic basis for favoring alcoholysis over hydrolysis in water. Unlike hydrolases that perform alcoholysis only under anhydrous conditions, MsAcT demonstrates alcoholysis in substantially aqueous media and, in
L Sisfontes et al.
Clinica chimica acta; international journal of clinical chemistry, 155(2), 117-122 (1986-03-16)
Patients attending a general hospital for various reasons were screened for raised serum gamma glutamyltransferase (gamma-GT) and positive blood alcohol concentration (BAC). The results served as objective biochemical tests of heavy drinking. Among 419 individuals, 50 (11.9%) met these requirements

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service