Skip to Content
Merck
All Photos(1)

Key Documents

39430

Sigma-Aldrich

N,N-Dimethylaniline

puriss. p.a., ≥99.5% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N(CH3)2
CAS Number:
Molecular Weight:
121.18
Beilstein:
507140
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

puriss. p.a.

Assay

≥99.5% (GC)

impurities

aniline and monomethyl aniline, in accordance
substances darkened by HCl, in accordance

refractive index

n20/D 1.557 (lit.)
n20/D 1.558

bp

193-194 °C (lit.)

mp

1.5-2.5 °C (lit.)

density

0.956 g/mL at 25 °C (lit.)

cation traces

Al: ≤5 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤1 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤50 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤100 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

SMILES string

CN(C)c1ccccc1

InChI

1S/C8H11N/c1-9(2)8-6-4-3-5-7-8/h3-7H,1-2H3

InChI key

JLTDJTHDQAWBAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marius Koch et al.
Journal of the American Chemical Society, 134(8), 3729-3736 (2012-01-31)
The fluorescence quenching of 3-cyanoperylene upon electron transfer from N,N-dimethylaniline in three room-temperature ionic liquids (RTILs) and in binary solvent mixtures of identical viscosity has been investigated using steady-state and time-resolved fluorescence spectroscopy. This study was stimulated by previous reports
Yong Wang et al.
The journal of physical chemistry. B, 114(8), 2964-2970 (2010-02-12)
This paper addresses the experimentally observed mechanistic differences between the cytochrome P450-catalyzed N-demethylation of substituted N,N-dimethylanilines (DMA) and of N,N-dimethylbenzamides (DMBA). The two reactions of these substrates are initiated by C-H activation of the methyl groups on the nitrogen. Thus
Cai-Rong Zhang et al.
Journal of molecular graphics & modelling, 38, 419-429 (2012-11-03)
The photon to current conversion efficiency of dye-sensitized solar cells (DSCs) can be significantly affected by dye sensitizers. The design of novel dye sensitizers with good performance in DSCs depend on the dye's information about electronic structures and optical properties.
Haijun Li et al.
Organic letters, 11(18), 4176-4179 (2009-08-18)
Methylene-bridged bis-1,3-dicarbonyl derivatives were synthesized efficiently by iron-catalyzed oxidative reactions of 1,3-dicarbonyl compounds and N,N-dimethylaniline. Bipyrazoles and substituted 1,4-dihydropyridine were obtained by the reactions of bis-1,3-dicarbonyl compounds with hydrazines and ammonium acetate, respectively.
Hohjai Lee et al.
Nano letters, 11(12), 5367-5372 (2011-11-03)
We used a fluorescent chemical indicator of magnetic field to visualize the magnetic field around ferromagnetic nanostructures. The indicator was a chain-linked electron donor-acceptor molecule, phenanthrene-(CH2)12-O-(CH2)2-dimethylaniline, that forms spin-correlated radical pairs upon photoexcitation. The magnetic field altered the coherent spin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service