Skip to Content
Merck
All Photos(1)

Documents

390216

Sigma-Aldrich

2-Bromo-1,1,3-trimethoxypropane

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH(Br)CH(OCH3)2
CAS Number:
Molecular Weight:
213.07
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

95%

refractive index

n20/D 1.458 (lit.)

bp

54-55 °C/21 mmHg (lit.)

density

1.395 g/mL at 25 °C (lit.)

SMILES string

COCC(Br)C(OC)OC

InChI

1S/C6H13BrO3/c1-8-4-5(7)6(9-2)10-3/h5-6H,4H2,1-3H3

InChI key

GAFSXURUCDQGFB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Gas-chromatographic measurement of the level of dimethylacetal of alpha-bromine-beta-methoxypropionic aldehyde in the air of the work area].
Z F Kos'ianova et al.
Gigiena i sanitariia, (1)(1), 74-75 (1991-01-01)
Yu Wang et al.
Chemical communications (Cambridge, England), (8)(8), 1090-1091 (2005-02-19)
Using sodium deoxycholate as a protective medium, the selective recognition of Cu(II) at ng ml(-1) level is realized through dynamic phosphorescence quenching of 1-bromo-2-methylnaphthalene (BMN) without deoxygenation. The limit of detection is 4.32 ng ml(-1), and the relative standard deviation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service