Skip to Content
Merck
All Photos(1)

Key Documents

36893

Supelco

trans-Permethrin solution

10 μg/mL in cyclohexane, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20Cl2O3
CAS Number:
Molecular Weight:
391.29
Beilstein:
4153590
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in cyclohexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

CC1(C)[C@H](\C=C(\Cl)Cl)[C@H]1C(=O)OCc2cccc(Oc3ccccc3)c2

InChI

1S/C21H20Cl2O3/c1-21(2)17(12-18(22)23)19(21)20(24)25-13-14-7-6-10-16(11-14)26-15-8-4-3-5-9-15/h3-12,17,19H,13H2,1-2H3/t17-,19+/m1/s1

InChI key

RLLPVAHGXHCWKJ-MJGOQNOKSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-0.4 °F

Flash Point(C)

-18 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamad Goldust et al.
Annals of parasitology, 59(1), 31-35 (2013-07-09)
Scabies is a common parasitic infestation that is an important public-health problem in many resource-poor regions. It is commonly treated with the insecticides but the treatment of choice is still controversial. This study aimed at comparing the efficacy of permethrin
Ting Yang et al.
Journal of medical entomology, 50(3), 585-593 (2013-06-28)
Insecticides, especially pyrethroids, are important components in the vector-control effort. To better understand the development of resistance, the current study characterized resistance profiles in individual single-egg-raft colonies of a field population of Culex quinquefasciatus Say, HAmCq(G0). Our study, with 104
Sarah Gray et al.
The New Zealand medical journal, 126(1373), 53-61 (2013-06-26)
To assess the acceptability and feasibility of delivering targeted primary health care in a decile one primary school setting. A pilot public health nurse (PHN)-led clinic was set up in a South Auckland primary school (roll approximately 400). The clinic
Juan Du et al.
Environmental toxicology and chemistry, 32(6), 1403-1411 (2013-02-21)
Compared with acute toxicity, chronic exposures to low levels of contaminants are more environmentally relevant, but fewer data are available. In the present study, sediment toxicity of the pyrethoid permethrin to Chironomus dilutus was determined. The whole-life-cycle toxicity testing was
Raweewan Srisawat et al.
The Southeast Asian journal of tropical medicine and public health, 43(6), 1380-1386 (2013-02-19)
One of the mechanisms responsible for pyrethroid resistance in mosquitoes is mutations in domain IIS6 of voltage-gated sodium channel gene (kdr). Aedes aegypti larvae were collected from the central provinces of Thailand (Bangkok, Prachin Buri and Ratchaburi) and colonized until

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service