Skip to Content
Merck
All Photos(1)

Key Documents

36672

Sigma-Aldrich

N,N-Dicyclohexylmethylamine

purum, ≥97.0% (GC)

Synonym(s):

N-Methyldicyclohexylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)2NCH3
CAS Number:
Molecular Weight:
195.34
Beilstein:
2074991
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥97.0% (GC)

refractive index

n20/D 1.49 (lit.)
n20/D 1.491

bp

265 °C (lit.)

density

0.912 g/mL at 25 °C (lit.)

SMILES string

CN(C1CCCCC1)C2CCCCC2

InChI

1S/C13H25N/c1-14(12-8-4-2-5-9-12)13-10-6-3-7-11-13/h12-13H,2-11H2,1H3

InChI key

GSCCALZHGUWNJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

may discolor to brown on storage

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service