Skip to Content
Merck
All Photos(1)

Key Documents

35914

Supelco

Chlordimeform hydrochloride

PESTANAL®, analytical standard

Synonym(s):

Chlorodimeform hydrochloride, Chlorophenamidine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13ClN2 · HCl
CAS Number:
Molecular Weight:
233.14
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

Cl.CN(C)\C=N\c1ccc(Cl)cc1C

InChI

1S/C10H13ClN2.ClH/c1-8-6-9(11)4-5-10(8)12-7-13(2)3;/h4-7H,1-3H3;1H/b12-7+;

InChI key

WZFNZVJGDCKNME-RRAJOLSVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlordimeform hydrochloride is a broad spectrum insecticide and acaricide, which can be applied to the foliage of fruits, field crops and vegetables in order to control insects and mites.[1] It can also be effectively used against cattle ticks, since it can act like an anti-parasiticidal and antifeedant agent.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paaranjape, K. et al.
The Pesticide Encyclopedia (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service