Skip to Content
Merck
All Photos(1)

Key Documents

34775

Sigma-Aldrich

2,6-Di-tert-butyl-4-methylpyridine

purum, ≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H23N
CAS Number:
Molecular Weight:
205.34
Beilstein:
130503
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

purum

Assay

≥97.0% (GC)

color

brown

refractive index

n20/D 1.4763 (lit.)

bp

233 °C (lit.)

mp

33-36 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1cc(nc(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C14H23N/c1-10-8-11(13(2,3)4)15-12(9-10)14(5,6)7/h8-9H,1-7H3

InChI key

HVHZEKKZMFRULH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Physical form

filled as molten mass

Other Notes

Hindered base for glycosidations following a modified Koenigs-Knorr reaction [1][2][3]; Base for the conversion of carbonyls in vinyl triflates with triflic anhydrides [4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D. Crich, S. Sun
The Journal of Organic Chemistry, 62, 1198-1198 (1987)
W.J. Scott et al.
Organic Syntheses, 68, 116-116 (1990)
G. Desmares et al.
Helvetica Chimica Acta, 84, 880-880 (2001)
F.W. Lichtenthaler et al.
European Journal of Organic Chemistry, 3849-3849 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service