34164
Bis(2-ethylhexyl)phthalate-3,4,5,6-d4
analytical standard
Synonym(s):
DEHP-3,4,5,6-d4, Deuterated DEHP, Deuterated bis(2-ethylhexyl)phthalate, Dioctyl phthalate-3,4,5,6-d4, Phthalic acid-3,4,5,6-d4 bis(2-ethylhexyl ester)
About This Item
Recommended Products
grade
analytical standard
Quality Level
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
bp
231 °C/5 mmHg (lit.)
mp
-50 °C (lit.)
application(s)
agriculture
cleaning products
cosmetics
food and beverages
personal care
format
neat
mass shift
M+4
SMILES string
[2H]c1c([2H])c([2H])c(C(=O)OCC(CC)CCCC)c(c1[2H])C(=O)OCC(CC)CCCC
InChI
1S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20H,5-10,13-14,17-18H2,1-4H3/i11D,12D,15D,16D
InChI key
BJQHLKABXJIVAM-SAQXESPHSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service