Skip to Content
Merck
All Photos(1)

Key Documents

33480

Sigma-Aldrich

1,4-Diazabicyclo[2.2.2]octane

purum, ≥95.0% (NT)

Synonym(s):

TED, Triethylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2
CAS Number:
Molecular Weight:
112.17
Beilstein:
103618
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor pressure

2.9 mmHg ( 50 °C)

grade

purum

Assay

≥95.0% (NT)

refractive index

n20/D 1.4634 (lit.)

mp

156-159 °C (lit.)
156-159 °C

density

1.02 g/mL at 25 °C (lit.)

SMILES string

N12CCN(CC2)CC1

InChI

1S/C6H12N2/c1-2-8-5-3-7(1)4-6-8/h1-6H2

InChI key

IMNIMPAHZVJRPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Hoffmann et al.
PloS one, 4(10), e7644-e7644 (2009-11-06)
Mammalian ATAD3 is a mitochondrial protein, which is thought to play an important role in nucleoid organization. However, its exact function is still unresolved. Here, we characterize the Caenorhabditis elegans (C. elegans) ATAD3 homologue (ATAD-3) and investigate its importance for
György Stuber et al.
Molecular cancer, 8, 23-23 (2009-03-28)
The low molecular weight compound, PRIMA-1MET restores the transcriptional transactivation function of certain p53 mutants in tumor cells. We have previously shown that PRIMA-1MET induces nucleolar translocation of p53, PML, CBP and Hsp70. The Epstein-Barr virus encoded, latency associated antigen
Céline Ricour et al.
The Journal of general virology, 90(Pt 1), 177-186 (2008-12-18)
Theiler's murine encephalomyelitis virus (TMEV or Theiler's virus) is a neurotropic picornavirus that can persist lifelong in the central nervous system of infected mice, causing a chronic inflammatory demyelinating disease. The leader (L) protein of the virus is an important
Frauke Beilstein et al.
Journal of virology, 83(17), 8938-8945 (2009-06-26)
The UL33 protein of herpes simplex virus type 1 (HSV-1) is thought to be a component of the terminase complex that mediates the cleavage and packaging of viral DNA. In this study we describe the generation and characterization of a
Xuezhi Cui et al.
Brain and behavior, 1(2), 73-86 (2012-03-09)
Mutations in the L1 gene cause severe brain malformations and mental retardation. We investigated the potential roles of L1 in the regulation of choline acetyltransferase (ChAT) and in the development of septal cholinergic neurons, which are known to project to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service