Key Documents
27172
Sigma-Aldrich
Chromyl chloride on silica-alumina
extent of labeling: ~0.5 mmol/g CrO2Cl2 loading
About This Item
UNSPSC Code:
12352103
extent of labeling
~0.5 mmol/g CrO2Cl2 loading
loss
≤1% loss on drying
Other Notes
Selective supported oxidant, useful for the oxidation of alcohols into aldehydes and ketones
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
J.S. Filippo, jr. et al.
The Journal of Organic Chemistry, 42, 2182-2182 (1977)