Skip to Content
Merck
All Photos(1)

Documents

24260

Sigma-Aldrich

Chlorodicyclohexylborane solution

purum, ~1 M in hexane

Synonym(s):

Dicyclohexylboron chloride, Dicyclohexylboryl chloride, Dicyclohexylchloroborane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)2BCl
CAS Number:
Molecular Weight:
212.57
Beilstein:
3233942
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

grade

purum

reaction suitability

reagent type: reductant

concentration

~1 M in hexane

density

0.74 g/mL at 20 °C

SMILES string

ClB(C1CCCCC1)C2CCCCC2

InChI

1S/C12H22BCl/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h11-12H,1-10H2

InChI key

PBBOKJIYEZCTEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Aldol addition reactions
  • Mukaiyama aldol addition
  • Microwave assisted ring closing metathesis reactions
  • C-alkylation of aromatic aldimines
  • Reversible ketone-ketone aldol reactions
  • Syn- and anti-stereoselective aldol reaction hexenone with aldehydes

Other Notes

Ideal reagent for the generation of E-enol borinates from ketones in high stereoselectivity

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service