20190
tert-Butyllithium solution
technical, ~1.7 M in pentane
Synonym(s):
Lithium-2-methyl-2-propanide, t-BuLi
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)3CLi
CAS Number:
Molecular Weight:
64.06
Beilstein:
3587204
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
Recommended Products
grade
technical
shelf life
limited shelf life
concentration
~1.7 M in pentane
density
0.65 g/mL at 20 °C
storage temp.
2-8°C
SMILES string
[Li]C(C)(C)C
InChI
1S/C4H9.Li/c1-4(2)3;/h1-3H3;
InChI key
BKDLGMUIXWPYGD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Shigeo Hayashi et al.
Journal of enzyme inhibition and medicinal chemistry, 29(6), 846-867 (2014-02-13)
Because of the pivotal role of cyclooxygenase (COX) in the inflammatory processes, non-steroidal anti-inflammatory drugs (NSAIDs) that suppress COX activities have been used clinically for the treatment of inflammatory diseases/syndromes; however, traditional NSAIDs exhibit serious side-effects such as gastrointestinal damage
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service