Skip to Content
Merck
All Photos(1)

Key Documents

20185

Sigma-Aldrich

sec-Butyllithium solution

purum, ~1.3 M in cyclohexane

Synonym(s):

Lithium-2-butanide, s-BuLi

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)Li
CAS Number:
Molecular Weight:
64.06
Beilstein:
3587206
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
Pricing and availability is not currently available.

grade

purum

shelf life

limited shelf life

concentration

~1.3 M in cyclohexane

density

0.769 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

[Li]C(C)CC

InChI

1S/C4H9.Li/c1-3-4-2;/h3H,4H2,1-2H3;

InChI key

VATDYQWILMGLEW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

during storage a white precipitate of lithium hydride may form

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service