Skip to Content
Merck
All Photos(1)

Key Documents

12758

Supelco

Tetraundecyl benzophenone-3,3′,4,4′-tetracarboxylate

Selectophore

Synonym(s):

ETH 2041, Tetraundecyl 4,4′-carbonyl-diphthalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C61H98O9
CAS Number:
Molecular Weight:
975.43
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
Pricing and availability is not currently available.

product line

Selectophore

SMILES string

CCCCCCCCCCCOC(=O)c1ccc(cc1C(=O)OCCCCCCCCCCC)C(=O)c2ccc(C(=O)OCCCCCCCCCCC)c(c2)C(=O)OCCCCCCCCCCC

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Plasticizer of high lipophilicity for clinically relevant ion-selective electrodes and multisensing field effect transistors (ISFET)[1]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U. Oesch et al.
Chimia, 40, 351-351 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service