Skip to Content
Merck
All Photos(1)

Key Documents

06837

SAFC

Benzamidine hydrochloride hydrate

Synonym(s):

Benzamidine hydrochloride hydrate, Amidinobenzene hydrochloride, Benzamidinium chloride, Benzenecarboximidamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(=NH)NH2 · HCl · xH2O
CAS Number:
Molecular Weight:
156.61 (anhydrous basis)
Beilstein:
3594959
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Assay

≥99.0% (HPLC)

form

powder or crystals

concentration

98.0-102.0 % (w/w) (anhydrous basis, AT)
98.0-102.0 % (w/w) (anhydrous basis, NT)

impurities

≤20 mg/kg heavy metals
≤3000 mg/kg residual solvent (methanol) (GLC-HS)
≤5000 mg/kg residual solvents (2-propanol, acetone) (GLC-HS) (each)
8-19% water

mp

86-88 °C (lit.)

solubility

H2O: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

Cl[H].[H]O[H].NC(=N)c1ccccc1

InChI

1S/C7H8N2.ClH.H2O/c8-7(9)6-4-2-1-3-5-6;;/h1-5H,(H3,8,9);1H;1H2

InChI key

BQVJCMMLDJPBFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Benzamidine HCl is a reversible inhibitor of trypsin, trypsin-like enzymes, and serine proteases. A concentration of approximately 1 mM is used for general protease inhibition. To inhibit proteases from yeast, a range of 0.5 to 4.0 mM is used and it is for the most part interchangeable with pepstatin A.

In addition to being a strong competitive inhibitor of trypsin, benzamidine HCl has been also shown to be a strong competitive inhibitor of thrombin and plasmin. It was also found to be as effective as aprotinin in the prevention of glucagon degradation in human plasma.
Peptidase inhibitor.

Preparation Note

Benzamidine HCl is soluble in water and alcohol. Solubility testing in water at a concentration of 50 mg/ml yields a clear solution with heating. Benzamidine HCl is sensitive to oxidation. It is recommended to prepare solutions fresh each time in degassed water prior to use. However, frozen aliquots stored under inert gas, to exclude air, may be stable for a short time. Insufficient information is available to assess the shelf-life of a frozen solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valeriy V Pak et al.
Cell metabolism, 31(3), 642-653 (2020-03-05)
Hydrogen peroxide (H2O2) is a key redox intermediate generated within cells. Existing probes for H2O2 have not solved the problem of detection of the ultra-low concentrations of the oxidant: these reporters are not sensitive enough, or pH-dependent, or insufficiently bright

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service