Skip to Content
Merck
All Photos(1)

Documents

06811

Supelco

Conductivity Standard

conductivity 200 μS/cm at 25 °C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

≤2.5% KCl (T)

conductivity

198.0-202.0 μS/cm at 25 °C

density

0.995-1.014 g/cm3 at 20 °C

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

single component solution

Looking for similar products? Visit Product Comparison Guide

Application

Conductivity calibration solution to monitor and verify conductivity measurement devices.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - STOT RE 2 Oral

Target Organs

Liver,Kidney

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Temperature and concentration dependence of the ionic transport properties of lithium-ion battery electrolytes
Landesfeind J and Gasteiger HA
Journal of the Electrochemical Society, 166(14), A3079-A3079 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service