Skip to Content
Merck
All Photos(1)

Key Documents

06703

Sigma-Aldrich

O,O′-Bis(2-aminoethyl)octadecaethylene glycol

≥95% (oligomer purity)

Synonym(s):

PEG-diamine (n=20)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H84N2O19
CAS Number:
Molecular Weight:
897.10
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95% (oligomer purity)

reaction suitability

reagent type: cross-linking reagent

SMILES string

NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCN

InChI

1S/C40H84N2O19/c41-1-3-43-5-7-45-9-11-47-13-15-49-17-19-51-21-23-53-25-27-55-29-31-57-33-35-59-37-39-61-40-38-60-36-34-58-32-30-56-28-26-54-24-22-52-20-18-50-16-14-48-12-10-46-8-6-44-4-2-42/h1-42H2

InChI key

PEPHTALHHSCONG-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service