Skip to Content
Merck
All Photos(1)

Key Documents

02677

Sigma-Aldrich

Boc-N-Me-D-Leu-OH

≥98.0%

Synonym(s):

Boc-N-methyl-D-leucine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(COOH)N(CH3)COOC(CH3)3
CAS Number:
Molecular Weight:
245.32
Beilstein:
4184551
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0%

form

powder or crystals

reaction suitability

reaction type: Boc solid-phase peptide synthesis

color

white to light yellow

application(s)

peptide synthesis

SMILES string

CC(C)C[C@@H](N(C)C(=O)OC(C)(C)C)C(O)=O

InChI

1S/C12H23NO4/c1-8(2)7-9(10(14)15)13(6)11(16)17-12(3,4)5/h8-9H,7H2,1-6H3,(H,14,15)/t9-/m1/s1

InChI key

YXJFAOXATCRIKU-SECBINFHSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service