Skip to Content
Merck
All Photos(1)

Key Documents

00416

Sigma-Aldrich

(S)-2-(Boc-amino)-1,3-diphenyl-1-propanone

purum, ≥96.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23NO3
CAS Number:
Molecular Weight:
325.40
Beilstein:
8002474
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥96.0% (HPLC)

form

powder

color

beige

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](Cc1ccccc1)C(=O)c2ccccc2

InChI

1S/C20H23NO3/c1-20(2,3)24-19(23)21-17(14-15-10-6-4-7-11-15)18(22)16-12-8-5-9-13-16/h4-13,17H,14H2,1-3H3,(H,21,23)/t17-/m0/s1

InChI key

ACUYZILXZGMZSY-KRWDZBQOSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service