Skip to Content
Merck
All Photos(1)

Documents

00400585

(−)-Bornyl acetate

primary reference standard

Synonym(s):

endo-(1S)-1,7,7-Trimethylbicyclo[2.2.1]hept-2-yl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20O2
CAS Number:
Molecular Weight:
196.29
Beilstein:
2502036
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

vapor pressure

0.6 mmHg ( 20 °C)

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

refractive index

n20/D 1.463 (lit.)

bp

223-224 °C (lit.)

density

0.986 g/mL at 20 °C (lit.)

storage temp.

−20°C

SMILES string

CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C

InChI

1S/C12H20O2/c1-8(13)14-10-7-9-5-6-12(10,4)11(9,2)3/h9-10H,5-7H2,1-4H3/t9-,10+,12+/m0/s1

InChI key

KGEKLUUHTZCSIP-HOSYDEDBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference standard in the analysis of herbal medicinal products.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

185.0 - 194.0 °F

Flash Point(C)

85 - 90 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service