Skip to Content
Merck
All Photos(1)

Key Documents

00045

Sigma-Aldrich

Boc-11-Aun-OH

≥98.0% (TLC)

Synonym(s):

11-(Boc-amino)undecanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H31NO4
CAS Number:
Molecular Weight:
301.42
Beilstein:
1975656
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (TLC)

form

powder with small lumps

reaction suitability

reaction type: Boc solid-phase peptide synthesis

color

white

mp

66-72 °C

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)NCCCCCCCCCCC(O)=O

InChI

1S/C16H31NO4/c1-16(2,3)21-15(20)17-13-11-9-7-5-4-6-8-10-12-14(18)19/h4-13H2,1-3H3,(H,17,20)(H,18,19)

InChI key

HPTPZJBSQUULAV-UHFFFAOYSA-N

Other Notes

Protected amino alkyl spacer; Reagent for introducing a peptidomimetic analogue of a tetrapeptide residue

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rafaela Cristina Sanfelice et al.
The journal of physical chemistry. B, 124(32), 7037-7045 (2020-07-18)
The synergistic use of blends of regioregular poly(3-hexylthiophene) (P3HT) and poly(vinylidene fluoride) (PVDF) or poly((vinylidene fluoride)-block-(methyl methacrylate)) (PVDF-PMMA) to form Langmuir and Langmuir-Schaefer (LS) films is reported. P3HT has wide applications in sensor devices because of its properties such as
S R Nagarajan et al.
Journal of medicinal chemistry, 40(10), 1422-1438 (1997-05-09)
MyristoylCoA:protein N-myristoyltransferase (NMT) covalently attaches the 14-carbon saturated fatty acid myristate, via an amide bond, to the N-terminal glycine residues of a variety of cellular proteins. Genetic studies have shown that NMT is essential for the viability of the principal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service