Skip to Content
Merck
All Photos(1)

Documents

40-0103

Sigma-Aldrich

pH standard solution

pH 6.86 (25 °C)

Synonym(s):

pH 6.86 Buffer Calibration Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700

availability

available only in Japan

pH

6.86 (25 °C)

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

Application


  • Machine learning-enhanced drug testing: A study employed machine learning techniques to enhance the detection of morphine and methaDonein urinary biofluids, utilizing solutions including pH standard solutions to maintain sample integrity during analysis (Habibi et al., 2024).

  • Synthesis of magnetic covalent organic frameworks: A study on the synthesis of amino-modified magnetic covalent organic frameworks for dye extraction in beverages utilized pH standard solutions to optimize the reaction conditions, highlighting their importance in ensuring reaction efficiency and stability (Guan et al., 2024).

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service