Skip to Content
Merck
All Photos(1)

Key Documents

28-4380

Sigma-Aldrich

Sodium tartrate dibasic dihydrate

JIS special grade, ≥99.0%

Synonym(s):

L-(+)-Tartaric acid disodium salt, Disodium tartrate dihydrate, Sodium tartrate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4Na2O6 · 2H2O
CAS Number:
Molecular Weight:
230.08
Beilstein:
6121732
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
JIS special grade
form:
solid

grade

JIS special grade

Assay

≥99.0%

form

solid

availability

available only in Japan

pH

7.0-9.0 (20 °C, 50 g/L)

SMILES string

[Na+].[Na+].[H]O[H].[H]O[H].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

InChI

1S/C4H6O6.2Na.2H2O/c5-1(3(7)8)2(6)4(9)10;;;;/h1-2,5-6H,(H,7,8)(H,9,10);;;2*1H2/q;2*+1;;/p-2/t1-,2-;;;;/m1..../s1

InChI key

FGJLAJMGHXGFDE-DGFHWNFOSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service