Skip to Content
Merck
All Photos(1)

Key Documents

28-2970

Sigma-Aldrich

Sodium hydroxide

≥96.0%, suitable for amino acid analysis

Synonym(s):

‘Caustic soda’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
Assay:
≥96.0%
vapor pressure:
<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)
Pricing and availability is not currently available.

vapor density

>1 (vs air)

vapor pressure

<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)

Assay

≥96.0%

form

solid

availability

available only in Japan

mp

318 °C (lit.)

suitability

suitable for amino acid analysis

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Virginie Mariot et al.
Annals of neurology, 78(3), 387-400 (2015-05-29)
Facioscapulohumeral muscular dystrophy (FSHD) is linked to either contraction of D4Z4 repeats on chromosome 4 or to mutations in the SMCHD1 gene, both of which result in the aberrant expression of the transcription factor DUX4. However, it is still difficult
Konstantinos A Polyzos et al.
Cardiovascular research, 106(2), 295-302 (2015-03-10)
Atherosclerosis is a chronic inflammatory disease that is initiated by the retention and accumulation of low-density lipoprotein in the artery, leading to maladaptive response of cells from the immune system and vessel wall. Strong evidence implicates indoleamine 2,3-dioxygenase (IDO), the
C Berger et al.
Human reproduction (Oxford, England), 30(4), 800-811 (2015-03-06)
Does ulipristal acetate (UPA) used for emergency contraception (EC) interfere with the human embryo implantation process? UPA, at the dosage used for EC, does not affect human embryo implantation process, in vitro. A single pre-ovulatory dose of UPA (30 mg)
Valdeir Arantes et al.
Biotechnology for biofuels, 7, 87-87 (2014-07-01)
There is still considerable debate regarding the actual mechanism by which a "cellulase mixture" deconstructs cellulosic materials, with accessibility to the substrate at the microscopic level being one of the major restrictions that limits fast, complete cellulose hydrolysis. In the
Jesper Hjortnaes et al.
Advanced healthcare materials, 4(1), 121-130 (2014-06-25)
Three dimensional (3D) hydrogel platforms are powerful tools, providing controllable, physiologically relevant microenvironments that could aid in understanding how various environmental factors direct valvular interstitial cell (VIC) phenotype. Continuous activation of VICs and their transformation from quiescent fibroblast to activated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service