05-5840
Copper(II) carbonate basic
CP
Synonym(s):
Cupric carbonate basic
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
CP
composition
Cu, ≥50%
reaction suitability
reagent type: catalyst
core: copper
availability
available only in Japan
SMILES string
[Cu++].O[Cu]O.[O-]C([O-])=O
InChI
1S/CH2O3.2Cu.2H2O/c2-1(3)4;;;;/h(H2,2,3,4);;;2*1H2/q;2*+2;;/p-4
InChI key
ZMMDPCMYTCRWFF-UHFFFAOYSA-J
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service