Skip to Content
Merck
All Photos(1)

Key Documents

01-1900

Sigma-Aldrich

Agar

SAJ first grade

Synonym(s):

Agar-agar, Gum agar

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H18O9)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
50405901
Pricing and availability is not currently available.

grade

SAJ first grade

availability

available only in Japan

InChI

1S/C14H24O9/c1-5-8(16)13-11(7(21-5)4-20-13)23-14-10(18)12(19-2)9(17)6(3-15)22-14/h5-18H,3-4H2,1-2H3/t5?,6-,7?,8-,9+,10-,11?,12+,13+,14?/m1/s1

InChI key

GYYDPBCUIJTIBM-DYOGSRDZSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Queeny Wing-Han Yuen et al.
The open biomedical engineering journal, 5, 39-46 (2011-06-07)
Cervical lymph nodes are common sites of metastatic involvement in head and neck cancers. These lymph nodes are superficially located and palpation is a common practice for assessing nodal hardness and staging cancer which is, however, too subjective and with
Evaluation of Brilliance VRE agar for the detection of vancomycin-resistant enterococci in rectal swab specimens.
Gozde Ongut et al.
Journal of medical microbiology, 62(Pt 4), 661-662 (2013-01-29)
Jin Xu et al.
Ultrasonics, 53(4), 889-896 (2013-01-24)
Histotripsy uses high-intensity focused ultrasound pulses at low duty cycle to generate energetic bubble clouds inside tissue to fractionate a region. As a potential tumor treatment modality, this cavitation-based non-invasive technique has the advantages of easy monitoring and sharp borders.
Hidekazu Nishimura
Kansenshogaku zasshi. The Journal of the Japanese Association for Infectious Diseases, 86(6), 723-733 (2013-02-02)
Several Japanese companies sell electrical devices advertised as effective in inactivating viruses and killing bacteria by releasing special materials, e.g., Plasmacluster ions, Nanoe particle and minus ions, into the air. These companies claim that their devices killed bacteria on plates
Malika Gouali et al.
Journal of clinical microbiology, 51(3), 894-900 (2013-01-04)
The performance of CHROMagar STEC and CHROMagar STEC O104 (CHROMagar Microbiology, Paris, France) media for the detection of Shiga toxin-producing Escherichia coli (STEC) was assessed with 329 stool specimens collected over 14 months from patients with suspected STEC infections (June

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service