Skip to Content
Merck
All Photos(1)

Documents

XX6320115

Millipore

Power Supply, 115 V

fecal coliforms, fecal streptococci, microorganisms

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104424
NACRES:
NB.11

AC/DC input

115 V, 60 Hz

application(s)

cosmetics
environmental
food and beverages
microbiology

shipped in

ambient

General description

115 V rechargeable nickel-cadmium battery

Application

fecal coliforms, fecal streptococci, microorganisms

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service