Skip to Content
Merck
All Photos(1)

Key Documents

WP62RBDNA

Millipore

High Output Pump rebuild kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105101
eCl@ss:
32012303
NACRES:
NB.24

shipped in

ambient

General description

This item includes parts for catalog numbers WP6211560
This item is a kit composed of the following items:
(1) Diaphragm hold-down plate assembly, (1) Valve plate assembly, (1) Pump head, (1) Vacuum gauge, (1) Pressure gauge, (4) Head screws, (2) Handle screws’

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service