Skip to Content
Merck
All Photos(1)

Key Documents

SPK94000102

Millipore

ISOPAK VALVE 1.5" SINTER RING SEAL KIT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

Pricing and availability is not currently available.

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EHU023701EHU001651EHU023031
Ensembl | human accession no.

ENSG00000112210

Ensembl | human accession no.

ENSG00000080371

Ensembl | human accession no.

ENSG00000104388

Ensembl | human accession no.

ENSG00000118508

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

Gene Information

human ... RAB23(51715), RAB23(51715)

Gene Information

human ... RAB21(23011), RAB21(23011)

Gene Information

human ... RAB2A(5862), RAB2(5862)

Gene Information

human ... RAB32(10981), RAB32(10981)

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

NCBI accession no.

NM_016277

NCBI accession no.

NM_014999

NCBI accession no.

NM_002865

NCBI accession no.

NM_006834


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service