Skip to Content
Merck
All Photos(1)

Key Documents

SLM-242

Sigma-Aldrich

RPMI-1640/ F12 complete medium

With ʟ-glutamine and 10% FBS, liquid, sterile-filtered, suitable for cell cutlure

Synonym(s):

Complete Medium, Roswell Park Memorial Institute 1640 medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.71
Pricing and availability is not currently available.

Product Name

RPMI / F12 Complete Medium, with 2 mM L-Glut and 10% FBS,

form

liquid

Quality Level

technique(s)

cell culture | mammalian: suitable

General description

RPMI / F12 Complete Medium contains a 1:1 mixture of RPMI-1640 medium and Ham’s F12 medium, with 2 mM L-glutamine and 10% U.S. origin FBS. RPMI / F12 Complete Medium is a convenient sterile solution for the culture of primary mammalian cells as well as a variety of normal and transformed cells.

Quality

pH= 7.2-7.5
Osmolality= 270-312 mOsm/kg
Endotoxin= ≤2 EU/mL
Mycoplasma= Negative
Sterility Tested= Negative for Growth
Appearance= Orange/ Red Clear Liquid
Cell Line Tested= LC-2/ad Human lung adenocarcinoma

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service