Skip to Content
Merck
All Photos(1)

Documents

S8001M

Sigma-Aldrich

CpGenome Human Methylated DNA Standard Set

It is intended for use as a positive control in gene methylation studies, such as bisulfite conversion of DNA with the CpGenome Turbo Bisulfite Modification Kit.

Synonym(s):

CpGenome Methylated DNA, Human DNA Standards, Methylated DNA Standard Set

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.77

Quality Level

species reactivity

human

manufacturer/tradename

CpGenome
Upstate®

application(s)

genomic analysis

shipped in

dry ice

Related Categories

General description

The CpGenome Human Methylated DNA Standard is purified from HCT116 DKO cells that contain genetic knockouts of DNA methyltransferases, DNMT1 (-/-) and DNMT3b (-/-) , and have less than 5% methylated DNA. This Standard has been methylated enzymatically at all CpG dinucleotides by M. SssI methyltransferase. It is intended for use as a positive control in gene methylation studies, such as bisulfite conversion of DNA with the CpGenome Turbo Bisulfite Modification Kit (Cat. No. S7847). Bisulfite-modified DNA can be evaluated by methylation-specific PCR (MSP) technology using the CpG WIZ amplification kits.

Materials Provided:

One vial containing 5 µg (20 µL) of CpGenome Human Methylated DNA Standard at a concentration of 250 ng/µL.

Validation:

Methylation-specific PCR (MSP) was performed on the CpGenome Human Methylated DNA after bisulfite modification with the CpGenome Turbo Bisulfite kit (Cat. No. S7847). Three sets of primers from the CpG WIZ BRCA1 amplification kit (Cat. No. S7830) were used in this assay: the U primer set, which anneals to unmethylated bisulfite-modified DNA; M primers, which anneal to methylated bisulfite-modified sequences; and W primers, which anneal to unmethylated or methylated DNA that has not undergone bisulfite modification. Only the M primers generated product for the methylated DNA standard.

CpGenome and CpG WIZ are trademarks of Serologicals Corporation. CpG WIZ Methylation Products apply to technologies exclusively licensed from The Johns Hopkins University School of Medicine. Methylation-specific PCR (MSP) technology is covered under U.S. Patent # 5,786,146.

Application

Research Category
Epigenetics & Nuclear Function

Physical form

Liquid in buffer containing 10 mM Tris-HCl, 1 mM EDTA, pH 8.0

Storage and Stability

Recommended Storage: Stable at -20°C for up to 6 months from date of receipt.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service