Skip to Content
Merck
All Photos(1)

Documents

NA46

Sigma-Aldrich

Anti-p300 Mouse mAb (NM-11)

liquid, clone NM-11, Calbiochem®

Synonym(s):

Anti-p300 antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

NM-11, monoclonal

form

liquid

does not contain

preservative

species reactivity

human, rat

should not react with

mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EP300(2033)

General description

Purified mouse monoclonal antibody generated by immunizing mice with the specified immunogen and fusing splenocytes with Sp2/0 mouse myeloma cells. Recognizes the p300 protein.
Recognizes the p300 protein in A431, HeLa, and ML-1 cells.
This Anti-p300 Mouse mAb (NM-11) is validated for use in Frozen Sections, Immunoblotting, Immunofluorescence, Immunoprecipitation, Paraffin Sections for the detection of p300.

Immunogen

Epitope: within amino acids 2071-2091 near the CBP C-terminus
Human
recombinant, human p300 protein

Application

Frozen Sections (see application references)

Immunoblotting (1 µg/ml)

Immunofluorescence (1-5 µg/ml)

Immunoprecipitation (1 µg/sample)

Paraffin Sections (1-5 µg/ml, heat pre-treatment required)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 0.05 M sodium phosphate buffer, 50% glycerol.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
A431, HeLa, or ML-1 cells

Other Notes

Antibody should be titrated for optimal results in individual systems.
Dallas, P.B., et al. 1997. Hybridoma16, 273.
Shisler, J., et al. 1996. J. Virol.70, 68.
Wang, H.G., et al. 1995. J. Virol.69, 7917.
Banerjee, A.C.,et al. 1994. Oncogene9, 1733.
Eckner, R., et al. 1994. Genes Dev.8, 869.
Abraham, S.E., et al. 1993. Oncogene8, 1639.
Wang, H.G., et al. 1993. J. Virol.67, 476.
Howe, J.A., and Bayley, S.T., 1992. Virology186, 15.
Rikitake, Y., and Moran, E., 1992. Mol. Cell. Biol.11, 2826.
Yaciuk, P., and Moran, W., 1991. Mol. Cell. Biol.11, 5389.
Egan, C., et al. 1988. Mol. Cell. Biol.8, 3955.
Yee, S. and Branton, P., 1985. Virology147, 142.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Stauffer et al.
The Journal of biological chemistry, 282(13), 9678-9687 (2007-02-03)
The highly related acetyltransferases, p300 and CREB-binding protein (CBP) are coactivators of signal-responsive transcriptional activation. In addition, recent evidence suggests that p300/CBP also interacts directly with complexes that mediate DNA replication and repair. In this report, we show that loss
Kevin M Elias et al.
JCI insight, 1(13) (2016-09-13)
Regulation of lineage-restricted transcription factors has been shown to influence malignant transformation in several types of cancer. Whether similar mechanisms are involved in ovarian cancer pathogenesis is unknown. PAX8 is a nuclear transcription factor that controls the embryologic development of
Karthigayan Shanmugasundaram et al.
Nature communications, 8(1), 997-997 (2017-10-21)
The molecular mechanisms that couple glycolysis to cancer drug resistance remain unclear. Here we identify an ATP-binding motif within the NADPH oxidase isoform, NOX4, and show that ATP directly binds and negatively regulates NOX4 activity. We find that NOX4 localizes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service