MABX8409
Anti-COVID-19, Nucleoprotein Antibody, 1068
suitable for ELISA
Synonym(s):
SARS-Cov-2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
1068, monoclonal
storage condition
avoid repeated freeze/thaw cycles
technique(s)
ELISA: suitable
isotype
IgG
shipped in
wet ice
General description
This mouse monoclonal Anti-COVID-19, Nucleoprotein, 1068, Cat. No. MABX8409 is tested for use in ELISA, for the detection of COVID-19 Nucleoprotein.
Specificity
COVID-19 & Coronavirus Nucleoprotein (NP)
Immunogen
Native Coronavirus
Application
Recommend MABX8410 for a paired immunoassay.
Target description
Coronavirus disease 2019 (COVID-19) is an infectious disease caused by the virus strain severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), a virus closely related to the SARS virus. A SARS-CoV-2 nucleoprotein antibody can be used to support the detection of SARS-CoV-2 (COVID-19, 2019-nCoV).
Physical form
Formulated with 10mM Phosphate Buffered Saline with 0.02% sodium azide, pH 7.0
Reconstitution
Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Storage and Stability
Short Term: 2-8°C. Long Term: -20°C. Avoid repeated freezing and thawing.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service