MABT199
Anti-Integrin Beta1 Antibody, clone 8E3
clone 8E3, from mouse
Synonym(s):
Integrin beta-1, Fibronectin receptor subunit beta, VLA-4 subunit beta, CD29
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
8E3, monoclonal
species reactivity
human
technique(s)
activity assay: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... ITGB1(3688)
General description
Specificity
Immunogen
Application
Cell Structure
ECM Proteins
Activity Assay Analysis: A representative lot from an independent laboratory stimulated cellular adhesion and increased ligand binding activity of Integrin Beta1 (Mould, A. P., et al. (2004). J Biol Chem. 280(56):4238-4226.).
Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated Integrin Beta1 in IP (Tiwari, S., et al. (2011). J Cell Sci. 124(10):1672-1680.).
Quality
Western Blot Analysis: A 1:2,000 dilution of this antibody detected Integrin Beta1 in 10 µg of HT-1080 cell lysate.
Target description
Physical form
Storage and Stability
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Analysis Note
HT-1080 cell lysate
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
recommended
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service