Skip to Content
Merck
All Photos(4)

Key Documents

MABS2321

Sigma-Aldrich

Anti-Biotin Antibody, clone 8H6.8

Synonym(s):

Vitamin B7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

8H6.8, monoclonal

purified by

using protein G

species reactivity (predicted by homology)

all

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

storage temp.

2-8°C

Specificity

Clone 8H6.8 is a mouse monoclonal antibody that detects biotin-conjugated proteins.

Immunogen

Biotin conjugated to KLH.

Application

Quality Control Testing

Evaluated by ELISA with Biotin-BSA.

ELISA Analysis: Serial dilutions from a representative lot detected Biotin-BSA in ELISA application.

Tested Applications

Western Blotting Analysis: A 1:5,000 dilution from a representative lot detected Biotin-BSA.

Immunofluorescence Analysis: A 1:50 dilution from a representative lot detected Biotin-conjugates in Human kidney tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Biotin, a bicyclic compound, is a water-soluble vitamin with a ureido (N-CO-N) and a tetrahydrothiophene ring. The ureido group is involved in its binding to avidin. The tetrahydrothiophene group is a valeric acid moiety. Although biotin molecule can exist in eight stereoisomers, the D-biotin isomer is the only one with biological activity. It plays important roles in a variety of critical metabolic reactions in the cell, and thus, is essential for normal human health, growth, and development. Its deficiency, particularly during pregnancy, leads to embryonic growth retardation, congenital malformation, and death. In mammals, biotin serves as a cofactor for four carboxylases that catalyze the transfer of a carboxyl group to targeted substrates. The four biotin-dependent carboxylases are: pyruvate carboxylase; propionly-CoA carboxylase; β-methylcrotonyl-CoA carboxylase; and acetyl-CoA carboxylase. These carboxylases play a critical role in the intermediate metabolism of gluconeogenesis, fatty acid synthesis, and amino acid catabolism. It is also reported to play a role in normal immune functions, including the production of antibodies, normal macrophage function, and the differentiation of T and B lymphocytes. This monoclonal antibody recognizes biotin conjugated to various proteins in ELISA and other immunochemical applications. (Ref.: Said, SM. (2012). Subcell. Biochem. 56; 1-19; Rabin, BS. (1983). J. Nutr. 113(11); 2316 2322).

Physical form

Purified mouse monoclonal antibody IgG1 in PBS with 0.05% sodium azide.

Reconstitution

1 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service