Skip to Content
Merck
All Photos(1)

Key Documents

MABS2214

Sigma-Aldrich

Anti-Extrahepatic biliary cells Antibody, clone hGB8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

hGB8, monoclonal

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgMκ

epitope sequence

Unknown

shipped in

ambient

target post-translational modification

unmodified

General description

The gallbladder is a small, distensible pouch-like organ that concentrates and stores bile produced by the liver that helps break down dietary fats. Bile flows out of the liver via hepatic ducts that join with the cystic duct, which conveys bile to and from the gall bladder. Gallbladder is derived from the foregut endoderm during embryonic development. In an un-distended state, its mucosa appears to be thrown into many folds. Its lumen is lined with a high columnar epithelium. This epithelial layer rests on a highly vascularized lamina propria. The biliary epithelium shows a morphological heterogeneity that is strictly associated with a variety of functions performed at the different levels of the biliary tree. These epithelial cells are also involved in absorption of water and inorganic salts and help to concentrate bile. The connective tissue wall of gallbladder contains abundant elastic fibers and layers of smooth muscles that run obliquely. Between the smooth muscle layers and serosa lies a thick layer subserosal layer of connective tissue. The availability of antibodies that preferentially mark cell populations within the extrahepatic biliary system are useful for identification and classification of gallbladder cells in relation to bile physiology, gallstone formation, cell plasticity and renewal, metaplasia, and tumor formation. Clone hBG8 is reported to detect human gallbladder mucosal epithelium cells with subcellular localization.(Ref.: Galivo, FH., et al. (2015). Stem Cell Res. 15(1); 172-181).

Specificity

Clone hGB8 is a mouse monoclonal antibody that detects subcellular mucosal epithelium in human gallbladder.

Immunogen

Extrahepatic human biliary cells

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in human gallbladder tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected mucosal epithelium with subcellular localization in human gallbladder tissue sections.

Tested Applications

Immunofluorescence Analysis: A representative lot detected Extrahepatic biliary cells in Immunofluorescence applications (Galivo, F.H., et al. (2015). Stem Cell Res. 15(1):172-81).

Immunohistochemistry (Paraffin) Analysis: A representative lot detected Extrahepatic biliary cells in Immunohistochemistry applications (Galivo, F.H., et al. (2015). Stem Cell Res. 15(1):172-81).

Flow Cytometry Analysis: A representative lot detected Extrahepatic biliary cells in Flow Cytometry applications (Galivo, F.H., et al. (2015). Stem Cell Res. 15(1):172-81).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Extrahepatic biliary cells, clone hGB8, Cat. No. MABS2214, is a mouse monoclonal antibody that detects subcellular mucosal epithelium in gallbladder and is tested in Flow Cytometry, Immunofluorescence, and Immunohistochemistry (Paraffin).

Physical form

Purified mouse monoclonal antibody IgM in PBS with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service