Skip to Content
Merck
All Photos(2)

Documents

MABN737

Sigma-Aldrich

Anti-OPA1, clone 1OPA-1A8 Antibody

ascites fluid, clone 10PA-1A8, from mouse

Synonym(s):

Dynamin-like 120 kDa protein, mitochondrial, Large GTP-binding protein, LargeG, Optic atrophy protein 1 homolog, Dynamin-like 120 kDa protein, form S1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

10PA-1A8, monoclonal

species reactivity

mouse, rat, human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... OPA1(4976)

General description

OPA1 (optic atrophy 1) is a dynamin-related GTPase that plays a role in genome preservation and mitochondrial morphology. Localization of OPA1 is thought to be anchored to the inner membrane of the mitochondria. OPA1 is highly expressed in inner and outer retinal neurons and is important for normal mitochondrial function in those cells. Mutations in OPA1 are the major cause of autosomal dominant optic atrophy. Recent research has found that OPA1 also has the ability to protect neurons from excitotoxic injury and could be a potential clinical target to increase endurance of neurons post brain damage.

Specificity

This antibody detects six known isoforms of OPA1 (Akepati, V. R., et al. (2008). J Neurochem. 106(1):372-383.).

Immunogen

Recombinant protein corresponding to mouse OPA1.

Application

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected OPA1 in COS cells (Prof. Mustapha Oulad-Abdelghani, IGBMC, France).

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated OPA1 from mouse brain tissue lysate (Akepati, V. R., et al. (2008). J Neurochem. 106(1):372-383.).
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
This Anti-OPA1 antibody, clone 1OPA-1A8 is validated for use in western blotting, ICC & IP for the detection of OPA1.

Quality

Evaluated by human brain tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected OPA1 in 10 µg of human brain tissue lysate.

Target description

~90 and ~100 kDa observed. Six isoforms between 80 kDa and 100 kDa are known to exist (Akepati, V. R., et al. (2008). J Neurochem. 106(1):372-383.).

Physical form

Mouse monoclonal IgG1κ ascites containing 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zaida Abad-Jiménez et al.
Antioxidants (Basel, Switzerland), 11(7) (2022-07-28)
The chronic low-grade inflammation widely associated with obesity can lead to a prooxidant status that triggers mitochondrial dysfunction. To date, Roux-en-Y gastric bypass (RYGB) is considered the most effective strategy for obese patients. However, little is known about its molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service