Skip to Content
Merck
All Photos(2)

Key Documents

MABN2254

Sigma-Aldrich

Anti-Catenin Delta-2 Antibody, clone 12B4

clone 12B4.1, from mouse

Synonym(s):

Catenin delta-2, GT24, Neural plakophilin-related ARM-repeat protein, NPRAP, Neurojungin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

12B4.1, monoclonal

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CTNND2(1501)

General description

Catenin delta-2 (UniProt: Q9UQB3; also known as Delta-catenin, GT24, Neural plakophilin-related ARM-repeat protein, NPRAP, Neurojungin) is encoded by the CTNND2 (also known as NPRAP) gene (Gene ID: 1501) in human. Delta Catenin is a component of the cadherin-catenin cell adhesion complex and is enriched in synaptosomes. Catenin Delta 2 is a member of the beta-catenin superfamily and is expressed in the brain where it is required for normal cognitive development and its highest expression is observed in fetal brain. Catenin Delta 2 is phosphorylated by Cdk5 and by GSK-3beta. Catenin Delta 2 plays a critical role in neuronal development, particularly in the formation and/or maintenance of dendritic spines and synapses. It is involved in the regulation of Wnt signaling where it acts on beta-catenin turnover, facilitating beta-catenin interaction with GSK3beta, phosphorylation, ubiquitination, and degradation. When bound to ZBTB33 (Zinc Finger and BTB Domain Containing 33), it functions as a transcriptional activator. The loss of CTNND2 gene has been associated with the intellectual disability observed in the cri du chat syndrome that is characterized by slow growth, microcephaly, delayed acquisition of psychomotor skills and intellectual disability. (Ref.: Yuan, L et al. (2015). J. Biol. Chem. 290(17):10947-57).

Specificity

Clone 12B4.1 specifically detects CTNND2 in rat brain microsomal preparations. This clone targets a sequeence within 104 amino acids in the C-terminal half of CTNND2.

Immunogen

GST/His-tagged recombinat fragment corresponding to 104 amino acids from the C-terminal half of human CTNND2.

Application

Anti-Catenin Delta-2 Antibody, clone 12B4, Cat. No. MABN2254, is a highly specific mouse monoclonal antibody that targets CTNND2 and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected Catenin Delta-2 in human cerebellum, human cerebral cortex, rat hippocampus, and rat cerebral cortex tissues.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in rat brain microsomal prep tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Catenin Delta-2 in 10 µg of rat brain microsomal preparations.

Target description

~136 kDa observed; 132.66 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service