MABN1853
Anti-Neuregulin-2 Antibody (NRG2), clone 8D11
clone 8D11, from mouse
Synonym(s):
Pro-neuregulin-2, membrane-bound isoform, Pro-NRG2, Neuregulin-2, NRG2, Divergent of neuregulin 1, DON-1
About This Item
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
8D11, monoclonal
species reactivity
rat, mouse, human
technique(s)
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
mouse ... Nrg2(100042150)
General description
Specificity
Immunogen
Application
Immunofluorescence Analysis: A representative lot detected NRG2 immunoreactivity among ErbB4+ interneurons in CA1 strata pyramidale and radiatum of 4% paraformaldehyde-fixed, 0.25% Triton X-100-permeabilized rat brain sections (Vullhorst, D., et al. (2015). Nat. Commun. 6:7222).
Neuroscience
Quality
Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected Purkinje cells surface Neuregulin-2 (NRG2) immunoreactivity in human cerebellum tissue sections.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service