Skip to Content
Merck
All Photos(1)

Documents

MABD390

Sigma-Aldrich

Anti-SPRY1 Antibody, clone 7E6.2

culture supernatant, clone 7E6.2, from mouse

Synonym(s):

Protein sprouty homolog 1, Spry-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

7E6.2, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SPRY1(10252)

General description

Protein sprouty homolog 1 (UniProt O43609; also known as Spry-1) is encoded by the SPRY1 gene (Gene ID 10252) in human. Sprouty was originally identified in Drosophila as a negative regulator of fibroblast growth factor (FGF) signaling during tracheal development. There exist four mammalian homologs (SPRY1-4) that likewise mediate feedback inhibition against growth factor-/receptor tyrosine kinase- (RTK-) dependent Ras/mitogen-activated protein (MAP) kinase signaling pathway. Loss of function studies in mice show that SPRY1 and SPRY2 play critical roles during early development of multiple organ rudiments, including external genitalia, eye, kidney, inner ear, tooth, and cortical patterning in the brain. In addition, evidences indicate that SPRY1 plays an antiproliferative and tumor-suppressing role in cancers. SPRY1 downregulation is reported in various cancer types, including prostate, breast, and ovarian cancers, and SPRY1 expresion has been proposed as a marker of good prognosis in patients with renal cell carcinoma.

Specificity

Clone 7E6.2 detects the ~35 kDa human SPRY1 by targeting an N-terminal region epitope.

Immunogen

GST-tagged recombinant human SPRY1 N-terminal fragment.

Application

Research Category
Stem Cell Research
This mouse monoclonal Anti-SPRY1 Antibody, clone 7E6.2, Cat. No. MABD390 is validated for use in Western Blotting for the detection of SPRY1.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected SPRY1 in 10 µg of SK-MEL-2 human skin melanoma cell lysate.

Quality

Evaluated by Western Blotting in HepG2 cell lysate.


Western Blotting Analysis: A 1:1,000 dilution of this hybridoma culture supernatant detected SPRY1 in 10 µg of HepG2 human hepatocellular carcinoma cell lysate.

Target description

~35 kDa observed. 35.12 kDa calculated.

Physical form

Mouse IgG1 hybridoma culture supernatant without azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service