Skip to Content
Merck
All Photos(1)

Key Documents

MAB1905

Sigma-Aldrich

Anti-Laminin α1/β 1 Antibody, clone AL-4

clone AL-4, Chemicon®, from rat

Synonym(s):

Laminin A Chain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

AL-4, monoclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Lama1(16772)

Specificity

As original characterized, clone AL-4 reacts with EHS mouse laminin, A chain (laminin α1). The epitope is mapped to the carboxy terminal globular domain on the A chain. Recent work by Scheele et al (2006) has further demonstrated that clone AL-4, under reducing conditions, recognized both the 200 kDa β1/γ1 band and the 400 kDa α1 band of mouse EHS laminin. However the affinity for the 400 kDa band was very low. AL-4 also demonstrated strong binding the Laminin β1. By ELISA the epitope for the AL-4 seemed to be located on the Laminin β1 chain, however the clone also recognizes the laminin α1 chain in immunoblots, thus AL-4 seems to show mixed cross reactivity to laminins α1 and β1, with the β1 reactivity being more prominent.

Immunogen

Laminin isolated from mouse EHS tumor.

Application

ELISA/RIA

Western blot

Immunoprecipitation

Immunohistochemistry. Reactive on tissue frozen in OCT at -70°C. Sections may be treated with hyaluronidase. Shows reactivity to paraffin embedded EHS tumor.

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins
This Anti-Laminin α1/β 1 Antibody, clone AL-4 is validated for use in ELISA, IP, WB, IH(P) for the detection of Laminin α1/β 1.

Physical form

Format: Purified
Purified from ascites by Protein G affinity chromatography. Purified immunoglobulin in 0.02M PB, 0.25M NaCl, pH=7.6, 0.1% sodium azide

Storage and Stability

Maintain between 2 and 8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interaction between fibronectin and ?1 integrin is essential for tooth development.
Saito, K; Fukumoto, E; Yamada, A; Yuasa, K; Yoshizaki, K; Iwamoto, T; Saito, M; Nakamura et al.
Testing null
E Klaffky et al.
Developmental biology, 239(1), 161-175 (2002-01-11)
For implantation and placentation to occur, mouse embryo trophoblast cells must penetrate the uterine stroma to make contact with maternal blood vessels. A major component of the uterine epithelial basement membrane and underlying stromal matrix with which they interact is
Jean-Baptiste Dupont et al.
Molecular therapy. Methods & clinical development, 2, 15010-15010 (2015-06-02)
Preclinical gene therapy strategies using recombinant adeno-associated virus (AAV) vectors in animal models of Duchenne muscular dystrophy have shown dramatic phenotype improvements, but long-lasting efficacy remains questionable. It is believed that in dystrophic muscles, transgene persistence is hampered, notably by
The extracellular matrix protein laminin ?2 regulates the maturation and function of the blood-brain barrier.
Menezes, MJ; McClenahan, FK; Leiton, CV; Aranmolate, A; Shan, X; Colognato, H
The Journal of Neuroscience null
Nozomu Takata et al.
Data in brief, 8, 465-469 (2016-07-01)
This article contains data related to the research article entitled "Specification of embryonic stem cell-derived tissues into eye fields by Wnt signaling using rostral diencephalic tissue-inducing culture" Sakakura (2016) [1]. Mouse embryonic stem cells (ESC) were used for the generation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service