Skip to Content
Merck
All Photos(1)

Key Documents

HTS030M

Sigma-Aldrich

ChemiSCREEN PTH Membrane Preparation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000

Pricing and availability is not currently available.

biological source

human

recombinant

expressed in Chem-1 cells

form

liquid

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

General description

Full-length human PTH1R cDNA
Parathyroid hormone (PTH) plays a critical role in mineral ion homeostasis, particularly in bone and kidney (Mannstadt et al., 1999; Gensure et al., 2005). A related peptide, parathyroid hormone-related peptide (PTHrP), plays an important role in skeletal development. Both peptides exert their biological actions by binding to a class B GPCR, PTH1R, that signals primarily by activation of adenylate cyclase (Schipani et al., 1993). Mutations in PTH1R have been identified as the cause of Jansen′s metaphyseal chondrodysplasia, Blomstrand′s chondrodysplasia, and enchondromatosis (Schipani and Provot, 2003). Intermittant treatment with PTH has important clinical utility in building bone mass in patients with osteoporosis (Rosen, 2003). Chemicon′s cloned human PTH1R-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant PTH1R expression on the cell surface and contains high levels of the promiscuous G protein Galpha15 to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for antagonists of interactions between PTH1R and its ligands.

Application

Human PTH1 GPCR membrane preparation for Radioligand binding Assays & GTPgammaS binding.
Radioligand binding assay, and GTPgammaS binding.

Biochem/physiol Actions

  • GPCR Class: B
  • Protein Target: PTH1
  • Target Sub-Family: PTH receptor

Quality

SPECIFICATIONS: Bmax: 2.66 pmol/mg
Kd: 0.75 nM

Specifications

RECOMMENDED ASSAY CONDITIONS: Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.

Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C

Radioligand: [125I] PTH (Perkin Elmer#: NEX397)

Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl , 0.1% BSA, filtered and stored at 4°C.

One vial contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 5-fold signal:background with 125I-labeled PTH at 0.75 nM

Physical form

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA no preservatives.Packaging method: Membranes protein were adjusted to 0.5 mg/ml in 1 ml packaging buffer, rapidly frozen, and stored at -80°C.

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service