HTS027M
ChemiSCREEN GnRH Membrane Preparation
Human GnRH / LHRH GPCR membrane preparation for Radioligand binding Assays & GTPgammaS binding.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.84
Recommended Products
biological source
human
Quality Level
recombinant
expressed in Chem-1 cells
manufacturer/tradename
ChemiScreen
Chemicon®
concentration
0.5 mg/mL
technique(s)
ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
General description
Gonadotropin-releasing hormone (GnRH), also known as luteinizing hormone-releasing hormone (LHRH), regulates the reproductive hormonal cascade in vertebrates. Upon release from the hypothalamus, GnRH stimulates secretion of luteinizing hormone and follicle-stimulating hormone from the pituitary. In humans, the type I GnRH receptor is a GPCR that is unusual in the lack of a cytoplasmic C-terminal tail. Although some species contain a type II GnRH receptor, the human version of this second receptor appears to contain mutations that render it inactive. GnRH analogs (agonists) are used in low doses in a pulsatile fashion in the treatment of infertility, delayed puberty and cryptorchidism. GnRH agonists at high doses desensitize the receptor, and are used along with antagonists in the treatment of hormone-dependent diseases (Millar et al., 2004). Chemicon′s cloned human type I GnRH receptor-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant GnRH receptor expression on the cell surface and contains high levels of the promiscuous G protein Galpha15 to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists and antagonists of the GnRH receptor.
Human Type I GnRH receptor
Application
Radioligand binding assay, and GTPgammaS binding.
Biochem/physiol Actions
GPCR Class: A
Protein Target: GnRH / LHRH
Target Sub-Family: GnRH
Quality
SPECIFICATIONS: Bmax: 1.0 pmol/mg
Kd: 0.3 nM
Kd: 0.3 nM
Specifications
Inucbation Conditions
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50 mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C
Radioligand: [125I]-[D-Trp6]-LHRH (Perkin Elmer # NEX365)
Wash Buffer: 50 mM Hepes, pH 7.4, 500 mM NaCl , 0.1% BSA, filtered and stored at 4°C.
One vial contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 3-fold signal:background with 125I-labeled [D-Trp6]-LHRH at 0.5nM.
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50 mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C
Radioligand: [125I]-[D-Trp6]-LHRH (Perkin Elmer # NEX365)
Wash Buffer: 50 mM Hepes, pH 7.4, 500 mM NaCl , 0.1% BSA, filtered and stored at 4°C.
One vial contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 3-fold signal:background with 125I-labeled [D-Trp6]-LHRH at 0.5nM.
Physical form
Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA no preservatives.Packaging method: Membranes protein were adjusted to 2.0 mg/ml in 1 mL packaging buffer, rapidly frozen, and stored at -80oC.
Storage and Stability
Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Gonadotropin-releasing hormone receptors.
Millar, Robert P, et al.
Endocrine Reviews, 25, 235-275 (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service