Skip to Content
Merck
All Photos(1)

Key Documents

FX0284

Supelco

Florisil® Adsorbent 60-100 Mesh

Meets Reagent Specifications for testing USP/NF monographs

Synonym(s):

Florisil® Adsorbent 60-100 Mesh, Magnesium Silicate Powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41115711
EC Index Number:
215-681-1

Quality Level

form

powder

color

white

particle size

150-250 μm
60-100 mesh (>= 80%)

pH

9.5 (20 °C, 100 g/L in H2O, slurry)

shipped in

ambient

storage temp.

room temp

Analysis Note

ColorWhiteDescriptionTo pass testFormPowderMesh size (60-100)80% min

Legal Information

Florisil is a registered trademark of U.S. Silica Company

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service