Skip to Content
Merck
All Photos(1)

Key Documents

EX0565

Supelco

Ethylene glycol

Synonym(s):

Ethylene Glycol, 1,2-Ethanediol, 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6O2
CAS Number:
Molecular Weight:
62.07
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-473-3

vapor pressure

0.053 hPa ( 20 °C)

Assay

≥98% (GC)

form

liquid

pH

6-7.5 (20 °C, 100 g/L in H2O)

bp

197.6 °C/1013 hPa

mp

-14--10 °C

transition temp

flash point 111 °C

density

1.11 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Application

  • Engineering Escherichia coli for the utilization of ethylene glycol: This study explores the potential of engineered E. coli to consume ethylene glycol, presenting a novel approach to biochemical production (Pandit et al., 2021).
  • Experimental study on thermal conductivity of ethylene glycol containing hybrid nano-additives: This research develops a new correlation for the thermal conductivity of ethylene glycol with nano-additives, enhancing its effectiveness in industrial applications (Afrand, 2017).
  • Boosting Selective Oxidation of Ethylene to Ethylene Glycol Assisted by In situ Generated H2O2 from O2 Electroreduction: This article discusses a catalytic method that uses mesoporous carbon to produce H2O2 which oxidizes ethylene into ethylene glycol, presenting a sustainable production method (Guan et al., 2023).

Analysis Note

Assay (GC): 98% min
Color (APHA): 20 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service