Skip to Content
Merck
All Photos(1)

Documents

EX0273

Supelco

Specially Denatured Alcohol 3A

200 Proof

Synonym(s):

Specially Denatured Alcohol 3A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6O
Molecular Weight:
46.07
UNSPSC Code:
12352104

Quality Level

form

liquid

composition

ethyl alcohol, 94.8-95.4 % (w/w)
methyl alcohol, 4.55-5.00 % (w/w)

concentration

200 proof in alcohol (denatured)

impurities

≤0.0025% Acidity (as acetic acid)

color

APHA: ≤10

shipped in

ambient

storage temp.

room temp

General description

200 Proof

Application

200 Proof

Analysis Note

Acidity (as acetic acid)0.0025% maxAssay: Ethyl alcohol, wt./wt. %94.8-95.4%Assay: Methyl alcohol, wt./wt. %4.55-5.00%Color (APHA)10APHA maxFusel oil constituentsNone detectedOdortypicalReducing substances (permanganate test)Passes testResidue after evaporation0.0025% maxSpecific Gravity at 60°F (15.56°C)0.7930-0.7950Suspended matterPasses testWater solubilityPasses testWater-insoluble matterPasses test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service