Skip to Content
Merck
All Photos(1)

Documents

CX0300

Supelco

Calcium Sulfate

Synonym(s):

Anhydrite

Sign Into View Organizational & Contract Pricing


About This Item

Assay

≥98% (complexometric)

Quality Level

form

powder

impurities

≤0.003% Heavy metals (as Pb)
≤0.003% Iron (Fe)
≤0.005% Nitrate (NO3)
≤0.01% Chloride (Cl)
≤0.1% Insoluble in dilute hydrochloric acid
≤0.5% Magnesium and alkali salts

color

white

mp

1450 °C

solubility

2.4 g/L

storage temp.

room temp

Application

anhydrous GR

Analysis Note

Assay (complexometric):98% min
Chloride (Cl):0.01% max
Color:White to Off-White
Form:Powder
Heavy metals (as Pb):0.003% max
Insoluble in dilute hydrochloric acid:0.1% max
Iron (Fe):0.003% max
Magnesium and alkali salts:0.5% max
Nitrate (NO3):0.005% max

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service