Skip to Content
Merck
All Photos(1)

Documents

CPAFS3PRB

Milli-Q

Kit: Progard® 2 Pack & Simfilter & Millex Vent Filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
JA.14

packaging

pkg of 1 kit

Quality Level

compatibility

for use with AFS®

shipped in

ambient

General description

The PROGARD® Pretreatment Pack has been designed to pretreat water by removing large particles, organic compounds and free chlorine from tap water. This protects the reverse osmosis (RO) membrane against mineral scaling, organic fouling and chlorine oxidation.
Benefits:
  • Optimal protection of reverse osmosis (RO) membrane
  • Ensures system lifetime and performance are optimized

Application

For AFS® 3/3D systems

Components

  • PR0G00002 Progard pretreatment
  • AFBCTFAFS bacteria filter and vent kit

Legal Information

AFS is a registered trademark of Merck KGaA, Darmstadt, Germany
PROGARD is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service